Suspended Solids Certified Reference Material
Catalog # ACCUWC-SOL-SS-1
Supplier: AccuStandard
CAS Number:
Specifications
- Pk:100 mL
- Description:Suspended solids certified reference material
- Storage conditions:Ambient (>5 °C)
- GHS signal:Danger
- Volume:100 mL