Order Entry
Export
Orders LinkContactUsLinkComponent
Hexadecene

Hexadecene

Glass bottle, 500 mL
Catalog # ROTH3834.1
Supplier:   Roth Carl
Hexadecene
Catalog # ROTH3834.1
Supplier:  Roth Carl
CAS Number:  26952-14-7
Hexadecene

Glass bottle, 500 mL

CAS Number:  26952-14-7
Price
99,60 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • CAS Number:26952-14-7
  • Formula:C₁₆H₃₂
  • Molecular Weight:224.43 g/mol
  • Boiling Pt:276 °C (1013 hPa)
  • Melting Pt:–5 °C
  • EINECS:248-131-4
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications