Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Butanediol
2,3-Butanediol
2,3-Butanediol
2,3-Butanediol
EHERC10861320
:
EHRENSTORFER
:
2,3-Butanediol
EHERC10861320
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
90.12 g/mol
:
12,01602
:
184 °C (1013 hPa)
:
18…20 °C
:
1 g/cm³ (20 °C)
:
Ambient
:
208-173-6
:
513-85-9
:
C₄H₁₀O₂
:
MFCD00004523