Order Entry
Export
ContactUsLinkComponent
1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%
1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%
Catalog # 20291.157
Supplier:  VWR Chemicals
CAS Number:  
undefined
1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%
Catalog # 20291.157
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

About this item

 :
 :364.35 g/mol
EINECS:207-582-7
 :GHS07
Storage Temperature:Ambient
CAS Number:87095-89-4
Formula:C₁₄H₂₂N₂O₈·1H₂O
MDL Number:MFCD00149243