Order Entry
Export
Orders LinkContactUsLinkComponent
 

Zinc

1
Catalog # SIALBCR327-1EA
Supplier:   Merck
Zinc
Catalog # SIALBCR327-1EA
Supplier:  Merck
CAS Number:  7440-66-6
CAS Number:  7440-66-6
Price
375,00 €
Log in for availability
1 * 1 items
Product Details & Documents
  • CAS Number:7440-66-6
  • Formula:Zn
  • MDL Number:MFCD00011291
  • Molecular Weight:65.39 g/mol
  • Merck Index:12,10255
  • Boiling Pt:908 °C (1013 hPa)
  • Melting Pt:420 °C
  • Density:7.14 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-175-3
Specifications

Specifications

  • Pk:
    1

Specifications