Order Entry
Export
ContactUsLinkComponent
Easy Na Sodium Analyser
Easy Na Sodium Analyser
Catalog # METN30060051
Supplier:  Mettler - Toledo
undefined
Easy Na Sodium Analyser
Catalog # METN30060051
Supplier:  Mettler - Toledo

Specifications

  • Communication interface:
    USB A (Printer or Balance), USB B (EasyDirect PC software)
  • Description:
    Easy Na sodium analyser
  • Electrical:
    100 - 240 V, ±10%, 50/60 Hz, 1,25 A
  • Overall dimensions:
    170×220×350 mm; 2650 g
  • Pk:
    1

Specifications

About this item

The Easy Na Sodium Analyser determines the sodium content in food and other products simply and quickly.

  • Easy to use
  • Smartphone-style apps design
  • One LongClick™ needed
  • Save money
  • No calibration necessary
  • Simple result data management

Reduce sample preparation time and effort by using safer and less expensive chemicals while, specifically and exclusively measuring sodium ions.

Replace the silver nitrate (AgNO₃) titrant normally used for chloride determination with the less expensive and safer sodium reagents. Due to the standard addition technique procedure, no calibration is necessary. Just carry out the automated sensor conditioning and immediately start the analysis. Connect Sodium analyser to EasyDirect™ titration software or to a USB-P25 dot-matrix printer.

Analyser is easy to use; simply add a sample and ionic strength adjustment (ISA) solution, and start the measurement. Smartphone-style apps design is used to controls all major functions. One LongClick™ is all that’s necessary to start the last analysis run.

Certifications: UL listed and CSA certified. IEC and EN certified.
Delivery: Standard delivery–Sodium analyser with analyser head, EasyStir and magnetic stir bar, 10 ml burette, bottle head for sodium standard, DX222-Na Sodium ISE, DX205-SC reference electrode and cable, tubing set, 110 V power adapter, EasyDirect data transfer software (trial), and application brochure.