Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Enzymes
Proteinase K
Print
Proteinase K
200 U, 1
Catalog # EDVO626
Supplier:
EDVOTEK
Proteinase K
Catalog # EDVO626
Supplier:
EDVOTEK
CAS Number:
Return to Top
Proteinase K
200 U, 1
Specifications
Jump to:
Specifications
CAS Number:
Price
27,50 €
Log in
for availability
1 * 1 items
-
+
Return Policy
Product Details & Documents
Specifications
Specifications
Pk:
1
Pk Info:
200 U
Cat. No.:
EDVO626
Storage conditions:
Freezer
Enzyme name:
Proteinase K
CAS No.:
39450-01-6
Specifications