Native Bovine Bovine Serum Albumin (from Serum), Sulfathiazol
Catalog # AATB50029
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Sulfathiazol
- Protein/peptide type:Native
- Source:Serum
- Species:Bovine
- Protein synonyms:BSA
- Protein/peptide name:Bovine Serum Albumin
Specifications
About this item
Sulfathiazol-BSA conjugate is used for developing anti-sulfathiazol antibodies that are typically used in an ELISA assay.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Caution: For research use only.