Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Estradiol
Catalog # AATB50554
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:5 mg
- Conjugation:Estradiol
- Protein/peptide type:Native
- Source:Serum
- Species:Megathura crenulata
- Protein synonyms:KLH
- Protein/peptide name:Keyhole limpet hemocyanin
Specifications
About this item
Estradiol is an estrogen steroid hormone and the major female sex hormone.
- High immunogenicity
- Useful for antibody development
- Estradiol test building block
Caution: For research use only.