Order Entry
Export
ContactUsLinkComponent
Native Horseradish Horseradish peroxidase (from Serum), Fentanyl
Native Horseradish Horseradish peroxidase (from Serum), Fentanyl
  AATB50564
 :  
undefined
Native Horseradish Horseradish peroxidase (from Serum), Fentanyl
  AATB50564
 :  

 

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Fentanyl
  • Protein/peptide type:
    Native
  • Source:
    Serum
  • Species:
    Horseradish
  • Protein synonyms:
    HRP
  • Protein/peptide name:
    Horseradish peroxidase

 

 

Fentanyl-HRP conjugate is an essential tool for developing fentanyl assays and tests.

  • High immunogenicity
  • Useful for antibody development
  • Fentanyl test building block
 : For research use only.