Order Entry
Export
ContactUsLinkComponent
 
Human Streptavidin, Xtra™ IF647
 
undefined
Human Streptavidin, Xtra™ IF647

 

  • Conjugation:
    Alexa Fluor® 647
  • Protein/peptide type:
    Native
  • Species:
    Human
  • Protein synonyms:
    Avidin
  • Protein/peptide name:
    Streptavidin