Phalloidin, XFD594
Catalog # AATB23158
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:300 Tests
- Conjugation:Alexa Fluor® 594
- Protein/peptide type:Native
- Source:Amanita phalloides
- Species:Amanita phalloides
- Protein synonyms:heptapeptide phallotoxin
- Protein/peptide name:Phalloidin
Specifications
About this item
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- Well-cited conjugate
- High affinity and selectivity
- In house quality control
Caution: For research use only.