Native Amanita phalloides Phalloidin (from Amanita phalloides), Alexa Fluor® 750
Catalog # AATB23130
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:300 Tests
- Conjugation:Alexa Fluor® 750
- Protein/peptide type:Native
- Source:Amanita phalloides
- Species:Amanita phalloides
- Protein synonyms:heptapeptide phallotoxin
- Protein/peptide name:Phalloidin
Specifications
About this item
This NIR fluorescent phalloidin conjugate selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
- Brighter signal
- High affinity and selectivity
- In house quality control
Caution: For research use only.