Order Entry
Export
ContactUsLinkComponent
 
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Fluorescein
  AATB23101
 :  
undefined
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Fluorescein
  AATB23101
 :  

 

  • Pack type:
    Vial
  • Pk:
    300 Tests
  • Conjugation:
    Fluorescein
  • Protein/peptide type:
    Native
  • Source:
    Eukaryotic cells
  • Species:
    Amanita phalloides
  • Protein synonyms:
    Amanita phalloides
  • Protein/peptide name:
    Phalloidin

 

 

This green fluorescent phalloidin conjugate (equivalent to FITC labeled phalloidin) selectively binds to F-actins.

  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
 : For research use only.