Order Entry
Export
ContactUsLinkComponent
Native Streptavidin (from Streptomyces avidinii), iFluor® 350
Native Streptavidin (from Streptomyces avidinii), iFluor® 350
Supplier:  AAT Bioquest
Certificates
undefined
Native Streptavidin (from Streptomyces avidinii), iFluor® 350
Supplier:  AAT Bioquest

Specifications

  • Conjugation:
    iFluor® 350
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin|Streptomyces avidinii
  • Protein/peptide name:
    Streptavidin

Specifications