Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), Alexa Fluor® 594
Catalog # AATB16892
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Alexa Fluor® 594
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin|Streptomyces avidinii
- Protein/peptide name:Streptavidin
Specifications
About this item
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- Extremely high biotin-binding affinity
- High quality conjugate
Caution: For research use only.