Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 647
Supplier: AAT Bioquest
Certificates
About this item
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
- Bright signal
- More photostable fluorescence
- Improved proprietary fluorescent label
Caution: For research use only.
Specifications
- Conjugation:iFluor® 647
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin|Streptomyces avidinii
- Protein/peptide name:Streptavidin