Order Entry
Export
ContactUsLinkComponent
Screen Quest™ Fluorimetric ELISA cAMP Assay Kit
Screen Quest™ Fluorimetric ELISA cAMP Assay Kit
Supplier:  AAT Bioquest
Certificates
undefined
Screen Quest™ Fluorimetric ELISA cAMP Assay Kit
Supplier:  AAT Bioquest

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Format:
    Pre-coated
  • Host:
    Human
  • Primary antibody reactivity:
    Human
  • Target protein:
    cAMP
  • Detection method:
    Fluorescence microplate reader

Specifications