ContactUsLinkComponent
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Catalog # AATB22328
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Catalog # AATB22328
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Tests per kit:
    200 Tests
Specifications
About this item
Provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay
  • alternative to anti-BrdU
  • extremely sensitive
  • deep red fluorescence
The resulted iFluor® 647-labeled DNA formed in cells is imaged with a Cy5 filter set.
Uses XdU which is incorporated into cellular DNA during DNA synthesis