ContactUsLinkComponent
Live or Dead™ Yeast Viability Assay Kit
Live or Dead™ Yeast Viability Assay Kit
Catalog # AATB22475
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Live or Dead™ Yeast Viability Assay Kit
Catalog # AATB22475
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Tests per kit:
    100 Tests
Specifications
About this item
Differentiates live yeast with intact membranes will stain fluorescent green, and dead yeast with damaged membranes will stain fluorescent red.
  • robust protocol
  • live yeast will glow green
  • dead yeast will glow red
This kit contains two ready-to-use nucleic acid stains: MycoLight™ Green JJ98, a replacement for SYTO™ 9, and propidium iodide.
Quantitatively distinguishes live and dead yeast within minutes using flow cytometry