Cell Meter™ Cell Viability Assay Kit *Green Fluorescence*
Catalog # AATB22786
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:500 Tests
- Description:Cell Meter™ Cell viability assay kit *Green fluorescence*
- Tests per kit:500 Tests
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
- Results in under 2 hours
Caution: For research use only.