ContactUsLinkComponent
Annexin V, PacBlue Conjugate
  AATB20089
 :  
undefined
Annexin V, PacBlue Conjugate
  AATB20089
 :  
  • Pk:
    100 Tests
  • Description:
    Annexin V, PacBlue conjugate
  • Tests per kit:
    100 Tests
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
  • High stability
  • In-house quality control
  • Multiple research applications
  • Optimised for violet laser
 : For research use only.