Order Entry
Export
ContactUsLinkComponent
CytoFix™ Red Mitochondrial Stain
CytoFix™ Red Mitochondrial Stain
Catalog # AATB23200
Supplier:  AAT Bioquest
CAS Number:  
undefined
CytoFix™ Red Mitochondrial Stain
Catalog # AATB23200
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    500 Tests
  • Description:
    CytoFix™ Red mitochondrial stain
  • Purity:
    95%
  • Tests per kit:
    500 Tests

Specifications

About this item

CytoFix™ Red mitochondrial stain is a dye that selectively stains mitochondria independent of mitochondrial membrane potential gradient.

  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Useful for Multicolor Imaging
  • Robust
  • Retained After Fixation
Caution: For research use only.