MitoLite™ Green EX488
Catalog # AATB22675
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:500 Tests
- Description:MitoLite™ Green
- Purity:95%
- Tests per kit:500 Tests
Specifications
About this item
MitoLite™ Green is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
- Mitochondria-Specific Dye
- Cell-Permeable
- Robust
Caution: For research use only.