Order Entry
Export
Orders LinkContactUsLinkComponent
CYTO-13

CYTO-13

CYTO-13, 0,5 mL
Catalog # AATB17553
Supplier:   AAT Bioquest
CYTO-13
Catalog # AATB17553
Supplier:  AAT Bioquest
CAS Number:  
CYTO-13
CYTO-13

CYTO-13, 0,5 mL

CAS Number:  
Price
313,00 €
Log in for availability
1 * 0,5 mL
Product Details & Documents

CYTO-13 is a fluorogenic, DNA-selective and cell-permeant dye for analyzing DNA content in living cells.

  • Cell-Permeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence

5 mM DMSO Solution

Caution: For research use only.

Specifications

Specifications

  • Pk:
    0,5 mL
  • Description:
    CYTO-13
  • Cat. No.:
    AATB17553
  • Purity:
    90%

Specifications