Order Entry
Export
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 350)
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 350)
Supplier:  AAT BIOQUEST
Certificates
undefined
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 350)
Supplier:  AAT BIOQUEST

Specifications

  • Catalog No:
  • AATB16770
  • AATB16520
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • iFluor® 350
  • iFluor® 350
  • Clone:
  • IgG
  • IgG
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG (H, L)
  • IgG (H, L)
  • Cross adsorption:
  • Min×Human
  • Min×Human

Specifications