Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 546)
AATB16457
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Clone:IgG
- Conjugation:iFluor® 546
- Host:Goat
- Isotype:IgG (H, L)
- Reactivity:Mouse
- Cross adsorption:—
- Size:200 µg
- Pk:200 µG
iFluor® 546 is a bright orange fluorescent dye.
- Bright signal
- Increased photostability
- Minimal quenching on proteins
: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 546
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse