Order Entry
Export
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy5®)
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy5®)
  AATB16879
undefined
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy5®)
  AATB16879

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Clone:
    IgG
  • Conjugation:
    Cy5®
  • Host:
    Goat
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Rabbit
  • Cross adsorption:
    Yes
  • Size:
    1 mg
  • Pack type:
    Vial
  • Pk:
    1 mg

 

 

AAT Bioquest's anti-rabbit secondary antibodies are affinity-purified antibodies with well-characterised specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity-purified
  • Bright signal
  • Cross-adsorbed

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit