Order Entry
Export
ContactUsLinkComponent
Anti-IgG (H, L) Goat Polyclonal Antibody (iFluor® 568)
Anti-IgG (H, L) Goat Polyclonal Antibody (iFluor® 568)
Supplier:  AAT Bioquest
Certificates
undefined
Anti-IgG (H, L) Goat Polyclonal Antibody (iFluor® 568)
Supplier:  AAT Bioquest

Specifications

  • Catalog No:
  • AATB16470
  • AATB16462
  • Antigen symbol:
  • IgG (H, L)
  • IgG (H, L)
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • iFluor® 568
  • iFluor® 568
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG (H, L)
  • IgG (H, L)

Specifications