0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Cyanine 3.5 azide
Cyanine 3.5 azide fluorescent probe
Cyanine 3.5 azide fluorescent probe
Irritant
Cyanine 3.5 azide fluorescent probe
Supplier:
AAT BIOQUEST
Certificates
Expand image
Irritant
Cyanine 3.5 azide fluorescent probe
Supplier:
AAT BIOQUEST
About this item
:
:
1073.11 g/mol
ADR:
5,II
:
GHS07