Order Entry
Export
ContactUsLinkComponent
Fluorescein aldehyde [5-FAM aldehyde] for biomolecular labeling
Irritant
Fluorescein aldehyde [5-FAM aldehyde] for biomolecular labeling
Catalog # AATB9000
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Fluorescein aldehyde [5-FAM aldehyde] for biomolecular labeling
Catalog # AATB9000
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :550.52 g/mol
ADR:5,II
 :GHS07