Order Entry
Export
ContactUsLinkComponent
Resorufin, sodium salt ≥90%
Irritant
Resorufin, sodium salt ≥90%
Catalog # AATB65
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Resorufin, sodium salt ≥90%
Catalog # AATB65
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :235.17 g/mol
ADR:5,II
 :GHS07
CAS Number:34994-50-8
Formula:C₁₂H₆NNaO₃
MDL Number:MFCD00005037