Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AMCA Alkyne
AMCA Alkyne ≥95%
AMCA Alkyne ≥95%
AATB507
:
AAT BIOQUEST
:
AMCA Alkyne ≥95%
AATB507
:
AAT BIOQUEST
:
Pk:
1 mg
Seq:
2070
:
:
270.28 g/mol