0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
AMCA succinimidyl ester
AMCA succinimidyl ester ≥90%
AMCA succinimidyl ester ≥90%
AATB502
:
AAT BIOQUEST
:
AMCA succinimidyl ester ≥90%
AATB502
:
AAT BIOQUEST
:
Pk:
10 mg
Seq:
2275
:
:
330.29 g/mol