
m-PEG24-Mal for biomolecular labeling
Catalog # AATB4525
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:1239.45 g/mol
ADR:5,II
:GHS07
CAS Number:88504-24-9
Formula:C₅₆H₁₀₆N₂O₂₇
MDL Number:MFCD11041152