Orders LinkContactUsLinkComponent
iFluor® 555, Tyramide
iFluor® 555, Tyramide
200
Catalog # AATB45105
Supplier:   AAT Bioquest
iFluor® 555, Tyramide
Catalog # AATB45105
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 555, Tyramide
iFluor® 555, Tyramide
200
CAS Number:  
Price
469,00 €
Log in for availability
1 * 200 items
Product Details & Documents
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
  • Increased fluorescence signal
  • Suitable for low-abundance targets
  • Compatible with all major fluorescence platforms
Caution: For research use only.
Specifications
Specifications
  • Pk:
    200
Specifications