
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
Catalog # AATB4507
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:266.21 g/mol
ADR:5,II
:GHS07
CAS Number:55750-62-4
Formula:C₁₁H₁₀N₂O₆
MDL Number:MFCD00043141