Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Difluororesorcinol ≥90%

2,4-Difluororesorcinol ≥90%

Plastic bottle, 1 g
Catalog # AATB4023
Irritant
Supplier:   AAT Bioquest
2,4-Difluororesorcinol ≥90%
Catalog # AATB4023
Supplier:  AAT Bioquest
CAS Number:  195136-71-1
2,4-Difluororesorcinol ≥90%
2,4-Difluororesorcinol ≥90%

Plastic bottle, 1 g

CAS Number:  195136-71-1
Price
235,00 €
Log in for availability
1 * 1 g
Product Details & Documents

2,4-Difluororesorcinol is a precursor that can be used for preparing fluorescent dyes such as fluorescein, coumarin and resorufin analogs.

Synonyms:  2,4-Difluorobenzene-1,3-diol
  • CAS Number:195136-71-1
  • Formula:C₆H₄F₂O₂
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g

Specifications