0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
N-FMOC-cadaverine
N-FMOC-cadaverine
N-FMOC-cadaverine
Irritant
N-FMOC-cadaverine
Supplier:
AAT BIOQUEST
Certificates
Expand image
Irritant
N-FMOC-cadaverine
Supplier:
AAT BIOQUEST
About this item
:
ADR:
5,II
:
GHS07