Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-FMOC-ethylenediamine
N-FMOC-ethylenediamine ≥95%
N-FMOC-ethylenediamine ≥95%
Irritant
N-FMOC-ethylenediamine ≥95%
Supplier:
AAT BIOQUEST
Certificates
Expand image
Irritant
N-FMOC-ethylenediamine ≥95%
Supplier:
AAT BIOQUEST
About this item
CAS Number:
166410-32-8
:
Formula:
C₁₇H₁₈N₂O₂
:
282.34 g/mol
ADR:
5,II
:
GHS07