Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-ROX glycine
6-ROX glycine solution 25 µM reference standard, for PCR
6-ROX glycine solution 25 µM reference standard, for PCR
6-ROX glycine solution 25 µM reference standard, for PCR
Catalog # AATB395
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
6-ROX glycine solution 25 µM reference standard, for PCR
Catalog # AATB395
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
5 mL
Seq:
2210
Specifications
About this item
Product Details
:
:
591.65 g/mol
Storage Temperature:
Freezer
Product Details