Order Entry
Export
ContactUsLinkComponent
 
Irritant
N-Succinimidyl 6-Biotinamidohexanoate ≥90%
Catalog # AATB3010
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
N-Succinimidyl 6-Biotinamidohexanoate ≥90%
Catalog # AATB3010
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :454.54 g/mol
ADR:5,II
 :GHS07
CAS Number:72040-63-2
Formula:C₂₀H₃₀N₄O₆S
MDL Number:MFCD00065502