Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Carboxy-4,7-dichlororhodamine 6G (5-dR6G)
5-Carboxy-4,7-dichlororhodamine 6G (5-dR6G) ≥95%
5-Carboxy-4,7-dichlororhodamine 6G (5-dR6G) ≥95%
AATB300
:
AAT BIOQUEST
:
5-Carboxy-4,7-dichlororhodamine 6G (5-dR6G) ≥95%
AATB300
:
AAT BIOQUEST
:
Pk:
25 mg
Seq:
2315
:
:
628.59 g/mol