Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PE-Cy5 Tandem
PE-Cy5 Tandem fluorescent probe
PE-Cy5 Tandem fluorescent probe
AATB2610
:
AAT Bioquest
:
PE-Cy5 Tandem fluorescent probe
AATB2610
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
240000 g/mol
:
5,II
:
GHS07