Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-JOE alkyne
6-JOE alkyne ≥95%
6-JOE alkyne ≥95%
AATB249
:
AAT BIOQUEST
:
6-JOE alkyne ≥95%
AATB249
:
AAT BIOQUEST
:
Pk:
5 mg
Seq:
2210
:
:
542.32 g/mol