Chrysamine G
Plastic bottle, 5 mg
Catalog # AATB23055
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Chrysamine G is a lipophilic, bis-salicylic acid analog of Congo Red that displays both high (Kd = 200 nM; Bmax = 1.13 moles per mole of Ab40) and low (Kd = 38.77 µM; Bmax = 23.10 moles per mole of Ab40) affinity binding sites for beta-amyloid (Ab) fibrils.
- CAS Number:6472-91-9
- Formula:C₂₆H₁₈N₄O₆
- MDL Number:MFCD07186299
- Molecular Weight:482.44 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:5 mg



