0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Tide Fluor™ 1 CPG
Tide Fluor™ 1 CPG 500 Å, Tide Fluor™ fluorescent probe
Tide Fluor™ 1 CPG 500 Å, Tide Fluor™ fluorescent probe
AATB2240
:
AAT BIOQUEST
:
Tide Fluor™ 1 CPG 500 Å, Tide Fluor™ fluorescent probe
AATB2240
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
:
:
5,II
:
GHS07