Order Entry
Export
ContactUsLinkComponent
3,3-Dipropyloxacarbocyanine iodide [DiOC3(3) iodide] ≥95%
3,3-Dipropyloxacarbocyanine iodide [DiOC3(3) iodide] ≥95%
Catalog # AATB22039
Supplier:  AAT Bioquest
CAS Number:  
undefined
3,3-Dipropyloxacarbocyanine iodide [DiOC3(3) iodide] ≥95%
Catalog # AATB22039
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :488.36 g/mol