Order Entry
Export
ContactUsLinkComponent
CytoTrace™ Orange CMTMR ≥90%, CytoTrace™
Irritant
CytoTrace™ Orange CMTMR ≥90%, CytoTrace™
Catalog # AATB22014
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
CytoTrace™ Orange CMTMR ≥90%, CytoTrace™
Catalog # AATB22014
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :554.04 g/mol
ADR:5,II
 :GHS07
CAS Number:323192-14-9
Formula:C₃₂H₂₈ClN₃O₄