Di-8-ANEPPS ≥95%
Catalog # AATB21497
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:592.87 g/mol
ADR:5,II
:GHS07
CAS Number:157134-53-7
Formula:C₃₆H₅₂N₂O₃S
MDL Number:MFCD03453608